3-in-1 Apparel Dispenser, Clear Acrylic, TrippNT
Supplier: TrippNT
50127
10012-632EA 797.9 USD InStock
10012-632
3-in-1 Apparel Dispenser, Clear Acrylic, TrippNT
PPE Dispensers
Acrylic dispenser have 3 large bins to sort and store protective equipment and apparel.
  • Wall mountable dispenser with 3 large compartments
  • White PVC and clear acrylic construction
  • Dispenses consumables from bottom
  • Loads from top with dust protecting lid
  • Keyhole installation
In stock and ready to ship. American made by TrippNT. Total replacement lifetime guarantee.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty