KODAK® Safelight Filters, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
74917-1A 74915-01 74919-13 74917-OA 74918-03 74918-13 74916-OC 74919-OA 74919-1A
100494-246EA 140.77 USD
100494-246 100494-218 100494-282 100494-250 100494-258 100494-222 100494-276 100494-262 100494-270 100494-226 100494-242 100494-290 100494-284
KODAK® Safelight Filters, Electron Microscopy Sciences
Lights Light Boxes
KODAK® Safelight filters are scientifically designed filters for safelight lamps.
OC/light amber - For contact and enlarging papers. Available in 51/2" Ø; 31/4×43/4"; 5×7"; 8×10" and 10×12".
1/red - For some blue-sensitive materials, most phototypesetting materials, KODAGRAPH Projection, and some KODAK LINAGRAPH papers. Available in 51/2" Ø; 5×7"; 8×10" and 10×12".
1A/light red - For slow orthochromatic materials. KODALITH and KODAGRAPH orthochromatic materials, and high resolution plates. Available in 51/2" Ø; 5×7"; 8×10" and 10×12".
2/dark red - For fast orthochromatic materials, someMore Product Information