Anti-GST Goat Polyclonal Antibody (RPE (R-Phycoerythrin))

Supplier: Columbia Biosciences
D5-1310
10065-642EA 563.61 USD
10065-642
Anti-GST Goat Polyclonal Antibody (RPE (R-Phycoerythrin))
Antibodies
Type: Secondary
Antigen: GST
Clonality: Polyclonal
Clone:
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR