Anti-TFAP2B Rabbit Polyclonal Antibody
Supplier: Prosci
27-112
10100-576EA 760.88 USD
10100-576
Anti-TFAP2B Rabbit Polyclonal Antibody
Antibodies
TFAP2B belongs to the AP-2 family which is developmentally regulated and have distinct overlapping functions in the regulation of many genes governing growth and differentiation. TFAP2B binds DNA as a dimmer and can form homodimers or heterodimers with other AP-2 family members. It may be a candidate for conferring susceptibility to type 2 didabetes. This gene encodes a member of the AP-2 family of transcription factors. AP-2 proteins form homo- or hetero-dimers with other AP-2 family members and bind specific DNA sequences. They are thought to stimulate cell proliferation and suppressMore Product Information