S-adenosylmethionine (SAM) (32mM), New England Biolabs
Supplier: W.u.G. Dickgießer GmbH
B9003S
101227-496EA 47 USD InStock
101227-496
S-adenosylmethionine (SAM) (32mM), New England Biolabs
Buffers Enzyme Buffers
S-adenosyl-methionine (SAM) is included with all of the DNA methylases available from New England Biolabs, Inc
As well as certain restriction enzymes for which it is required. The SAM is prepared in a solution of 0.005 M H2SO4 and 10% ETOH and filtered.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty