Cold Extractor, COEX, VELP Scientifica
Supplier: VELP SCIENTIFIC INC.
F30540204
10184-174EA 11349.54 USD
10184-174 10184-180 10184-186
Cold Extractor, COEX, VELP Scientifica
Extractors Automatic Extractor Systems
Cold extractor that performs rapid fat extraction directly in the same glass crucibles that are used by the FIWE Series for raw fiber analysis.
  • Precise Results
  • Versatile
  • Convenient
Precise Results–Reduces fat to below 1% improving analytical precision. No loss – reduced sample remains in same crucible for analysis.
Versatile–Immediate raw fiber extraction after fat extraction here. Processing of individual or several samples.
Convenient–Fatty components collected for easy disposal. All reagents can be added through the same channel.