Disposable Box, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
64412
102091-100PK 157.44 USD
102091-100
Disposable Box, Electron Microscopy Sciences
Boxes Waste Disposal Boxes
These boxes are used to dispose of empty ampoules, broken glass, empty vials, biohazard wastes, etc.
  • Colorful graphics for quick identification
  • Assembly instructions on each box
  • Folding design for shipping and storage
The outer box is made from corrugated cardboard, the inner one is a 2-mil polypropylene bag. When the box is full, the safety cap is pulled into place and the entire unit is ready for disposal or incineration. There are no replacement bags for these boxes.