Wooden Storage Box for Pin Mounts, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
76507
102091-320EA 159.2 USD InStock
102091-320
Wooden Storage Box for Pin Mounts, Electron Microscopy Sciences
Boxes Storage Boxes
EMS offers a high end wooden storage box for the storage of 12.7 mm (1/2") pin mounts.
The box can store up to 128 mounts in rubber foam with complete indexed sides.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty