Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Glass Bead Sterilizers, Electron Microscopy Sciences

Supplier: Electron Microscopy Sciences
66116-10 66116-20 66116-5 66117-10 66117-20 66117-3 66117-5
102095-928EA 1209.99 USD InStock
102095-928 102095-930 102095-934 102095-936 102095-938 102095-940 102095-942
Glass Bead Sterilizers, Electron Microscopy Sciences
Sterilizers Glass Bead Sterilizers
Glass beads are suggested to be replaced at least once in three months

These glass bead sterilizers are quick and easy to use and they are compact and light weight which makes them suitable for table tops or workbenches. They are used to sterilize instruments used in most laboratories and doctors' offices, dentists' offices, clinics, hospitals etc, killing  spores and bacteria within 10 seconds. All units are supplied with one filling of glass beads. Glass beads size ranging 1.5 – 2.0 mm diameter.

Glass beads are suggested to be replaced at least once in three months. Clean the glassMore Product Information
Order Now
 

ORDER

Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
Description Supplier No. VWR Catalog Number Unit Price Quantity
Glass bead sterilizers, Model BS1000 66116-10 102095-928 Each
Retrieving

Glass bead sterilizers, Model BS2000 66116-20 102095-930 Each
Retrieving

Glass bead sterilizers, Model BS500 66116-5 102095-934 Each
Retrieving

Glass Bead Replacement, Model BS1000 66117-10 102095-936 Each
Retrieving

Glass Bead Replacement, Model BS2000 66117-20 102095-938 Each
Retrieving

Glass Bead Replacement, Model BS300 66117-3 102095-940 Each
Retrieving

Glass Bead Replacement, Model BS500 66117-5 102095-942 Each
Retrieving

This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com

Generate Barcodes
Generate PDF Catalog Page
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty