Paraffin Tissue Microarrays, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
69130-15 69130-25 69130-45 69130-55 69130-10 69130-20 69130-40 69131-01 69131-02 69131-03 69131-04
101765-094EA 991.45 USD InStock
101765-094 101765-096 101765-098 101765-100 102096-518 102096-520 102096-522 102096-524 102096-526 102096-528 102096-530
Paraffin Tissue Microarrays, Electron Microscopy Sciences
Tissue Embedders
Paraffin tissue microarrays (TMAs) are becoming more common in the laboratories. The DVD instruction is accompanying with each kit, will step by step guide you to perform through the entire TMAs process. Arraymold features include:
  • Allow pathologist or researcher to view many different samples on the same single slide
  • Helpful in finding IHC markers for control or quality assurance from hundreds of samples
  • Staining markers will be more consistent when hundreds of samples are on one slide
  • Used less costly antibody
  • Small enough to be stored in a drawer
  • Rubber
More Product Information
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty