Anti-SPA-1/SIPA-1 Rabbit Polyclonal Antibody (HRP (Horseradish PE (Phycoerythrin)rOxidase))

Supplier: Bioss
BS-1602R-HRP
10323-472EA 804.08 USD
10323-472
Anti-SPA-1/SIPA-1 Rabbit Polyclonal Antibody (HRP (Horseradish PE (Phycoerythrin)rOxidase))
Antibodies
The product of this gene is a mitogen induced GTPase activating protein (GAP). It exhibits a specific GAP activity for Ras-related regulatory proteins Rap1 and Rap2, but not for Ran or other small GTPases. This protein may also hamper mitogen-induced cell cycle progression when abnormally or prematurely expressed. It is localized to the perinuclear region. Two alternatively spliced variants encoding the same isoform have been characterized to date.

Type: Primary
Antigen: SPA-1/SIPA-1
Clonality: Polyclonal
Clone:
Conjugation: HRP
Public Immunogen Range: 351-450/1042
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR