Pump 11 Elite Syringe Pumps, Harvard Apparatus
Supplier: Harvard Apparatus
70-4500
103301-796EA 3653.85 USD InStock
103301-796
Pump 11 Elite Syringe Pumps, Harvard Apparatus
Pumps Syringe Pumps
The Pump 11 Elite is an accurate, low flow syringe pumps designed for use in applications including mass spec calibration, drug and nutritional studies, reactor dosing, and electro-spinning
  • Easy to use with an LCD color touch screen and icon interface
  • Outstanding flow performance
  • Easily create, save and run simple to complex methods without a PC
  • Alphanumeric keypad for easy Method naming and recall
  • Adjustable linear force up to 35 lbs
  • Upgrade new versions of software remotely
These compact syringe pumps carry on the tradition as the premier workhorseMore Product Information
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty