AirChek Badges, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
64472-10 64472-50 64472-20 64472-30 64472-40
103306-268EA 786.36 USD InStock
103306-268 103306-274 103306-396 103306-398 103306-400
AirChek Badges, Electron Microscopy Sciences
Gas Detection Systems
Monitor vapor levels, simply and effectively.
  • Clips onto technicians clothing
  • Record of name, location, time
Airchek Badges provide an innovative, simple and effective system for monitoring vapor levels, including Formaldehyde, Glutaraldehyde, Xylene, Toluene, and Isopropanol. The badges provide continuous sampling for personal monitoring and allowing the measurements required under Government regulations. One badge provides continuous sampling over 8 hours. A second badge can be used to supply information for analysis of 15-minute peak exposureMore Product Information
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty