Anti-STN1 Rabbit Polyclonal Antibody

Supplier: Sino Biological
101695-T10-50 101695-T10-100 101695-T10-200
103640-118EA 142.96 USD
103640-118 103645-940 103645-942
Anti-STN1 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human OBFC1, and purified by antigen affinity chromatography.

Type: Primary
Antigen: OBFC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR