XYZ Goniometer Head Kits and Components, Rigaku
Supplier: Rigaku Reagents
1013156 1013157 1013158 1013163 1013159 1015055 1013162 1013161 1015228 1013160 1015229
103729-010EA 1050.08 USD
103729-010 103729-012 103729-014 103729-016 103729-058 103729-062 103729-064 103729-008 103729-052 103729-054 103729-056
XYZ Goniometer Head Kits and Components, Rigaku
Goniometers
XYZ Goniometer Heads feature a IUCr standard goniometer mount for compatibility with most goniometers.
Each goniometer head allows ±2.5mm adjustments in X and Y and ±5mm in Z.
Información de suministro: Goniometer head kits are supplied with a storage case, goniometer head key, and interchangeable parts to support sample mounting with either a magnetic mount or a Z support for use with capillary or brass pin mounts.
Order Now
SPECIFICATIONS
Environmentally Preferable