Redi-Fold Biopsy Paper Starter Kit, IMEB
Supplier: IMEB INC MS
RF-150
103783-060CS 164.62 USD
103783-060
Redi-Fold Biopsy Paper Starter Kit, IMEB
Paper
This kit includes a convenient pop-up dispenser that holds 6000 papers, keeping them conveniently accessible and dry.
  • Redi-Fold pop-up dispenser included
  • 6000 biopsy papers included
  • Papers measure 2.5×3.5"
White biopsy papers are easy to use for securing smaller specimens. Simply fold the paper around the specimen and place directly into processing cassette. Standard processing schedule can be followed with excellent results.