Anti-PCK2 Rabbit Polyclonal Antibody (Cy5.5®)

Supplier: Bioss
BS-5002R-CY5.5
10406-728EA 823.27 USD
10406-728
Anti-PCK2 Rabbit Polyclonal Antibody (Cy5.5®)
Antibodies
Catalyzes the conversion of oxaloacetate (OAA) to phosphoenolpyruvate (PEP), the rate-limiting step in the metabolic pathway that produces glucose from lactate and other precursors derived from the citric acid cycle.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PCK2
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR