NovaClean™ Floor Cleaner, Micronova
Supplier: Micronova
NC1-5G NC1-Q NC1-G NC10-G
68400-016EA 601.91 USD
68400-016 68400-011 68400-012 10769-418
NovaClean™ Floor Cleaner, Micronova
Cleaning Agents Detergents and Surface Cleansers
Concentrated, all-purpose cleanroom detergent is designed for use between disinfectant applications and for residue removal. Free of mobile ions, this formula is effective on most surfaces, including conductive flooring and semi conductor applications.
  • All-purpose cleanroom detergent
  • Ideal for pharmaceutical and biotech applications
  • Use NC10-G amine-free formula for semi-conductor applications
  • Anti-static and safe on conductive flooring
  • Yields 60-128 gallons (227-485 L) of cleaning solution per 1 gallon container
  • Filtered to 0.1 µm; free of mobile ions
More Product Information