Climate Chambers ICH Series, Memmert
Supplier: Memmert
ICH110115V ICH110230V ICH260115V ICH260230V ICH750115V ICH750230V
10861-360EA 15997.62 USD
10861-360 10861-362 10861-364 10861-366 10861-368 10861-370
Climate Chambers ICH Series, Memmert
Testing Chambers Climatic Chambers
Memmert stability chambers are constructed of stainless steel inside and out. The ICH L stability chamber will save energy and perform well for any project.
  • From temperatures of at least 10 up to +60 °C including air jacket heating system for gentle all-around heating
  • Humidity range 10 to 80% rh - or rh OFF
  • Illumination unit consisting of 3 fluorescent lights with cold white light (standard illuminant D65, 6.500K) and 2 UV-lamps in the spectral range of 320-400 nm
  • Programmable with internal data logging via included AtmoCONTROL Software
  • Available in 3
More Product Information