DNA Markers and Ladders
Supplier: R & D Systems GmbH
50461 50327 50328 50333 50323 50471 50322 50330
12001-502EA 396.42 USD
12001-502 12001-778 12001-780 12001-786 12001-498 12001-500 12001-504 12001-730 12001-732
DNA Markers and Ladders
Electrophoresis Reagents Molecular Weight Markers
Lonza DNA Ladders are precision manufactured to provide rapid and easy size determination of your DNA samples.
  • Ideal for PCR or cloning verification
  • Plasmid-free
Lonza SimplyLoad™ DNA ladders are ready-to-load directly on your gel to maximize your time.
Lonza’s standard DNA ladders are ready-to-dilute prior to loading on your gel. Plasmid-free to ensure minimal background.
For all of Lonza’s ladders, each band has an ideal 50% G-C content for rapid, precise sizing and sharp resolution. Our ladders are free from contaminating plasmid DNA so they have minimal background, giving you the perfect gel the first time, every time.
SimplyLoad™ DNA ladders are supplied at ready-to-load concentrations in TE Buffer and 1X Ficoll® sample loading buffer containing Bromophenol blue and Xylene cyanol FF so you can get straight to your results.