VWR® Prefilled Histology Containers, 10% Neutral Buffered Formalin (NBF)
Supplier: Azer Scientific Inc.
PFNBF-20 16004-124
16004-112CS 366.38 USD InStock
16004-112 16004-124
VWR® Prefilled Histology Containers, 10% Neutral Buffered Formalin (NBF)
Sample Containers General Purpose Containers
Containers are half-filled with 10% neutral buffered formalin, for the collection and transport of tissue samples.
  • High clarity virgin polypropylene container construction
  • Polyethylene screw caps included
  • 20–120 mL containers are 95kPa compliant
Each container is made of high clarity virgin polypropylene and includes a polyethylene screw cap
Small-sized prefilled formalin containers (20 mL and 40 mL) feature a click-top closure. The first click assures users the seal is fully engaged. The second click ensures 95kPa compliance. This click-top designMore Product Information
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty