Nalgene® Biohazardous Waste Containers, Thermo Scientific
Supplier: Thermo Fisher Scientific
6370-0005 6370-0004 6370-0015
16211-134EA 382.85 USD InStock
16211-134 16211-136 16211-138
Nalgene® Biohazardous Waste Containers, Thermo Scientific
Bins Waste Bins
Autoclavable polypropylene waste containers are ideal for secondary containment of biohazardous waste materials.
  • Convenient one-step sterilization, with no need to remove autoclavable bag
  • Port opening in cover allows convenient one-handed disposal of materials, limiting exposure to infectious waste
  • Holds standard-size autoclavable bags
  • Red universal biohazard symbol is molded in English and Spanish
  • Leakproof side and bottom construction
Containers comply with OSHA 29CFR 1910.1030 for use as protection against bloodborne pathogens.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty