Anti-HSP65 Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
ADI-SPA-882-E
200062-792EA 417.47 USD
200062-792
Anti-HSP65 Mouse Monoclonal Antibody
Antibodies
Hsp65 is a member of the Hsp60 family of heat shock proteins isolated from Mycobacterium bovis BCG. Hsp65 from M. bovis is identical to that of M. tuberculosis, and similar to that of M. leprae, two highly pathogenic strains of mycobacterium. Hsp65 is the immunodominant antigen during mycobacterial infection and vaccination, and has been linked to the development of autoimmune adjuvant arthritis in rats.

  • For Western Blot, in vitro Assay

Recognizes bacteria and bovine HSP65.

Type: Primary
Antigen: HSP65
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR