Chemware® PTFE Balls, SAINT GOBAIN PERFORMANCE PLASTIC
Supplier: Saint Gobain Performance Plastics
D1069326 D1069390
26397-431PK 89.18 USD
26397-431 26397-464
Chemware® PTFE Balls, SAINT GOBAIN PERFORMANCE PLASTIC
Beads Plastic Beads
Chemware® PTFE balls are mainly used in column packing and mixed beds
  • Minimize “bumping” during boiling
  • Resistant to most chemicals
  • Non-contaminating
They are inert and can be used up to 550 °F (288 °C).
Envase: 100 balls per package.