JACKSON SAFETY® V80 MONOGOGGLE™ 211 Protective Goggles, KIMBERLY-CLARK PROFESSIONAL®
Supplier: Ansell Healthcare
16669
33001-232CS 691.15 USD
33001-232 89130-968 89166-274 89166-272 33001-050
JACKSON SAFETY® V80 MONOGOGGLE™ 211 Protective Goggles, KIMBERLY-CLARK PROFESSIONAL®
Goggles
Patented indirect ventilation system provides increased airflow, minimizes condensation and fogging, and protects against splash.
  • Pliable plastic nosepiece for snug, comfortable seal
  • Can be worn over most prescription eyeglasses
  • Goggles provide 99.9% U.V. protection
Soft-frame design features a pantascopic angle to improve work area visibility.
Certificaciones: Meets ANSI Z87.1+ standards.