Trypan blue solution_SC 0,4% w/v PBS Stain_histology pH 7.5 ±0.5, Corning®

Supplier: Charles River Laboratories
Danger

25-900-CI
45000-717EA 27.06 USD
45000-717
Trypan blue solution_SC 0,4% w/v PBS Stain_histology pH 7.5 ±0.5, Corning®
Trypan Blue

Trypan Blue is the most common stain used in cell count and viability assays.


Viable cells exclude the dye, while nonviable cells absorb the dye and appear blue. Cells should be in suspension as single cells in buffered saline before counting. Trypan blue can be used in assays providing total cell count, viable cell cell or nonviable cell count alone. Ths solution is provided at the common concentration of 0.4% (w/v).

Formula: C₃₄H₂₄N₆Na₄O₁₄S₄
MW: 960.82 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003969
CAS Number: 72-57-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR