Basic Tectonics Model
Supplier: VWR
470004-622EA 170.29 USD
470004-622
Basic Tectonics Model
Models Geology Models
This Tectonics Model demonstrates the principle of superposition and the concepts of transgression, regression, and compression.
  • Powerful visual
  • Model provides dramatic visualization of faults
  • Length 11¾"
  • Width: 3¾"
  • Height: 7⅞"
  • Aligned to NGSS standards (MS/HS: ESS1, ESS2)
Students can construct a logical sequence of five sedimentary layers in a compression box then apply a convergent force with the plunger in the box to explore each concept.
The applied force causes the layers to fold and fault. It includes instructions and all materials.