Draw a Circuit Pen
Supplier: KIDDER MFG. CO. LTD
8035-CEPP
470165-028EA 25.65 USD
470165-028
Draw a Circuit Pen
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
This pen provides a great platform for discovering, playing with, repairing, and designing electronics.
  • Pen Length: 10 mL
  • Great Prototyping Tool for All Ages
  • For Use in a Wide Range of Applications 
  • Nontoxic, Solvent-Free, and Water-Soluble
  • Conductive-Adhesive Capabilities
Electric Paint can be used as a liquid wire to draw or print graphical circuits or even as a conductive adhesive that eliminates the need for soldering equipment. Electric Paint also comes in handy when repairing electronics such as PCBs or even old TV remotes. The paint canMore Product Information