Testis, Showing Spermatogenesis
Supplier: VWR
470183-698EA 10.95 USD
470183-698
Testis, Showing Spermatogenesis
Slides Prepared Slides
Extremely Thin, Plastic Embedded Tissue For Greater Viewing Clarity.
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Rodent. Details of meiotic division clearly shown.