Ward's® Mini Vortexer
Supplier: VWR International
470230-486
470230-486CS 3092.1 USD InStock
470230-486
Ward's® Mini Vortexer
Shakers and Mixers Vortex Mixers
Powerful, Mini Vortex Mixer.
  • Powerful vortexing for tubes up to 50 mL
  • Instant pressure activated operation
  • Compact mini size
  • 2 year warranty
Powerful is the best word to describe this mini vortexer. With a 4mm orbit and fixed speed of 2800 rpm, the Vornado instantly vortexes even the largest samples, included nearly full 50 mL tubes. Despite its powerful motor, the Vornado has a footprint of less than 4 x 4 inches, allowing it to fit on even the most crowded bench. Instant touch operation means that the motor is automatically activated with slightMore Product Information
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty