Adjustable Funnel Stand
Supplier: Wards
CH0689A
470343-566EA 23.75 USD
470343-566
Adjustable Funnel Stand
Support Stands
Crafted from polished wood, this stand with a 1.5" hole diameter and a height of 17" provides stability and convenience for various funnel applications.
  • Funnel stand featuring a sturdyhardwood construction that will stand up to the wear and tear of the lab
  • Height extends up to 18"
  • Base is approximately 7³/₄" by 4¹/₄"
  • Stand comes with clamping screws for adjusting the height
  • Designed for use in professional laboratories, but is an ideal choice for science classrooms
Funnel stand made from quality polished wood. Hole diameter is 1.5". Height of theMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable