Sodium Spoon
Supplier: Wards
CH0737
470343-578EA 5.1 USD
470343-578
Sodium Spoon
Spoons
The medium-sized sodium spoon is specifically designed for accurate and controlled dispensing of sodium, ensuring safe and efficient handling in experiments.
  • This spoon is for holding sodium under water
  • By compressing handles the sodium spoon opens to allow sodium to be placed in the caged spoon
  • Comprised of rust-proof wire stem and strong cage to safely enclose the sodium
  • Cage measures 15 mm (0.59") diameter
  • Over all Length 248 mm (9.75")