SP Bel-Art Silicone-Free Lens Cleaning Tissues, Bel-Art Products, a part of SP
Supplier: Bel-Art Products
F24841-0000
490005-582BX 29.5 USD InStock
490005-582
SP Bel-Art Silicone-Free Lens Cleaning Tissues, Bel-Art Products, a part of SP
Wipes Disposable Wipes and Towels Lens Papers
Use these high-quality, lint-free tissues to quickly and easily clean glass and plastic lenses.
  • Tissues do not contain silicone
  • Will not scratch glass or plastic lenses
  • 150 sheets packaged in a pop-up dispenser pack
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty