Benchmark BeadBlaster™ 24 Microtube Homogenizer
Supplier: Ward's
D2400
490005-842EA 16614.54 USD
490005-842
Benchmark BeadBlaster™ 24 Microtube Homogenizer
Homogenizers Benchtop Homogenizers
The BeadBlaster 24 completely lyses, grinds and homogenizes a broad variety of biological samples. Even the most difficult and resistant samples are fully homogenized, often in 35 seconds or less.
  • Fast, efficient homogenization in about 35 seconds
  • For all sample types, from soft tissue to bone
  • High throughput - up to 24 samples
  • Sealed, 2.0 mL tubes - no cross contamination
  • Cycle time: 1-90 sec, 1 sec increments
  • Memory: Maximum of 50 programs
  • Increment speed, 0.05, Pause between Cycles, 0-2 minutes, 1 sec increments
  • Warranty: 2-Years
The stainlessMore Product Information