SP Bel-Art Cryo-Safe™ Cold Box, Bel-Art Products, a part of SP
Supplier: Bel-Art Products
F18847-0002
490012-162EA 90.98 USD
490012-162
SP Bel-Art Cryo-Safe™ Cold Box, Bel-Art Products, a part of SP
Coolers Benchtop Coolers
Use this compact cooler on the benchtop to maintain twelve 1.5 mL centrifuge tubes at -15°C for 1.5 hours, or below 0°C for up to 4 hours eliminating ice baths or trips to the freezer.
  • Prevents expensive enzymes and critical samples from warming
  • Active by placing it in a -20°C freezer for 24 hours
  • Stackable
Units have a lid to protect the contents and can be stacked to save space during storage or transport.
Order Now
SPECIFICATIONS
Environmentally Preferable