VWR® Disposable Micropipettes
Supplier: VWR International
15401-402 15401-413 53432-706 53432-728 53432-740 53432-761 53432-783 53432-808 53432-921
15401-402CS 626.21 USD
15401-402 15401-413 53432-706 53432-728 53432-740 53432-761 53432-783 53432-808 53432-921
VWR® Disposable Micropipettes
Pipettes Microcapillaries
Borosilicate glass micropipettes are made to exacting specifications and are calibrated to contain with permanent fill lines.
  • Color-coded
  • Borosilicate glass fabrication
  • Sizes from 5 to 100 µl
  • Permanent fill lines
The 44.7 µl size is treated with the anticoagulant heparin for clinical applications.
Each canister is marked with bold size designations for quick identification in the laboratory.
Información para pedidos: This item is packed in easy dispensing canisters of 250 micropipettes and one 38.1 cm (15") aspirator tube.
Order Now
SPECIFICATIONS
Environmentally Preferable