Sonic Sifter Separator, 220 V
Supplier: ATM
L3P-26 L3P-25
57333-832EA 9025.19 USD InStock
57333-832 57333-834
Sonic Sifter Separator, 220 V
Shakers and Mixers Sieve Shakers
This superior laboratory sieving instrument is designed for fast, precise particle separations.
  • 220 V model
Sieving action can be varied for different densities and textures of materials. This unit utilizes a vertical oscillating column of air, which prevents particle attrition and screen wear, and a repetitive mechanical pulse, which helps reduce sieve binding and rearranges clustered particles. The Sonic Sifter Separator is easy to use with virtually maintenance free operation. Digital timer ensures repeatable results. Horizontal pulse accessoryMore Product Information
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty