SP Bel-Art 'C'-Shape Open Lead Ring Flask Weights with Vikem® Vinyl Coating, Bel-Art Products, a part of SP
Supplier: Bel-Art Products
F18308-0500 F18308-1000 F18308-2000 F18308-4000
74950-098EA 54.66 USD
74950-098 74950-100 74950-102 74950-104
SP Bel-Art 'C'-Shape Open Lead Ring Flask Weights with Vikem® Vinyl Coating, Bel-Art Products, a part of SP
Weights Flask Weights
These rings fit neatly around the neck of filtering flasks with side-arm tubing connections without disconnecting the tubing.
  • Green vinyl coating
  • Flexible
  • Available in four diameter/weight combinations
The open ring, made of lead and coated with green Vikem® vinyl to protect glassware, is flexible enough to be pulled apart to accommodate a wider vessel.