Alere iScreen® Saliva Alcohol Test Strips, Alere Toxicology
Supplier: Alere
I-SA-25
75780-118BX 67.89 USD
75780-118
Alere iScreen® Saliva Alcohol Test Strips, Alere Toxicology
Drug Tests
The iScreen® Saliva Alcohol Test Strip (Saliva) is a screening test used to semi-quantitatively measuring alcohol in human saliva. The test strip indicates relative Blood Alcohol Concentration (BAC) at 0.0%, 0.02%, 0.04%, 0.08%, and 0.30% cut-off levels.
The test is a firm plastic strip with a test pad attached at the tip. The test pad contains chemicals that are highly sensitive to alcohol. Alcohol in saliva reacts with the chemicals to produce a color change on the test pad. The color change depends on the amount of alcohol present. Results are read by comparing the color of the testMore Product Information