HRP Stripping Buffer, Azure Biosystems
Supplier: Fels-Chemie
AC2154
75794-868EA 227.51 USD InStock
75794-868
HRP Stripping Buffer, Azure Biosystems
Buffers Electrophoresis Buffers Stripping Buffers
HRP Stripping Buffer, Uniformly removes antibodies from developed HRP Western blots so the blot can be reprobed with another set of antibodies
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty