KIMBLE® Watch Glasses, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
72001-50 72001-70 72001-80 72001-90 72001-100 72001-125 72001-150
75797-130CS 31.84 USD
75797-130 75797-132 75797-134 75797-136 75797-138 75797-140 75797-142
KIMBLE® Watch Glasses, DWK Life Sciences
Watch Glasses
Concave circular glasses are used to perform evaporations for liquids, to cover beakers and to hold solids while weighing.
  • Minimum thickness 1.2 mm
  • Diameter tolerance ±1 mm
  • Fire polished edge
Manufactured from 90 expansion soda-lime glass.
Certificaciones: Conforming to USP Type III requirements.
Order Now
SPECIFICATIONS
Environmentally Preferable