Vibratory Sieve Shaker, AS 200 digital, Retsch
Supplier: Retsch
300300005 300310001
75804-638EA 3760.06 USD InStock
75804-638 75804-640
Vibratory Sieve Shaker, AS 200 digital, Retsch
Shakers and Mixers Sieve Shakers
These AS 200 series analytical sieve shakers work based on electromagnetic drive which produces a 3D throwing motion that moves the product to be sieved equally over the whole sieving surface
  • Controlled amplitude
  • Suitable for dry and wet sieving
  • Maintenance-free
  • Free adjustment of all process parameters
  • Easy operation, ergonomic design
This sieve system offeres excellent separation efficiency even with short sieving times. The most depicted feature of this sytem is that the digital display of vibration height and time. In this system, sieves can beMore Product Information
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty