2-Propanol 99% Antiseptic, Cleanser, Hydrox Laboratories

Supplier: NATIONAL DISTRIBUTION & CONTRACTING
Danger

HDX A0053
75830-308CS 282.12 USD
75830-308
2-Propanol 99% Antiseptic, Cleanser, Hydrox Laboratories
2-Propanol

  • Topical antiseptic
  • Active ingredient: isopropyl alcohol

Formula: (CH₃)₂CHOH
MW: 60.1 g/mol
Boiling Pt: 82
Melting Pt: -89
Flash Pt: 12 °C
MDL Number: MFCD00011674
CAS Number: 67-63-0
UN: 1219
ADR: 3 II
Merck Index: 13,05228
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ceac541820c6046c770c320cb35e80c1343901a5&vid=77f11bcf690db67b8cf7c1dbbc5d3a4d2586aeed&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR