RNase Erase™ Reagent, MP Biomedicals, LLC
Supplier: E. Becker & Co. GmbH
112440204
75856-412EA 146.2 USD InStock
75856-412
RNase Erase™ Reagent, MP Biomedicals, LLC
Cleaning Agents Decontaminants
Decontamination reagent for RNase.
  • Clear, colorless solution
  • Ready-to-use
  • Effectively removes RNA and RNase contamination
Spray on and rinse off for an RNase-free surface! Completely removes RNase contamination from bench tops, glass and plasticware. RNase Erase is nontoxic and comes packaged with an adjustable spray bottle for for ease of use.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty