Buck Ligature Scissors, OR-Grade, Sklar®
Supplier: Sklar
15-1359
76191-876EA 177.11 USD
76191-876
Buck Ligature Scissors, OR-Grade, Sklar®
Scissors
All Sklar products are industry standard.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar can deliver superior precision for any surgical procedure
Buck Ligature Scissors are used for removing ligatures. This product is straight with dual blunt tips, one of which is slightly hooked to slide under and gently lift up the ligature for cutting.