Folding Spill Containment Stand, Dynalon
Supplier: Dynalon
107374
76210-706EA 656.37 USD InStock
76210-706
Folding Spill Containment Stand, Dynalon
Trays/Dishes/Pots Spillage Trays/Pans
Folding Carboy Spill Stand allows undercounter access due to fold-away spill trough.
  • Manufactured of 0.375" high density polyethylene
  • With corrosion-resistant stainless steel hardware
Folding Carboy Spill Stand is designed for bench-top use, with a folding feature to allow access to drawers and other materials below. This heavy-duty spill tray will hold carboys up to 15" in diameter.
Main spill containment tray has a 2 gallon capacity and bottom spill trough will hold up to 1/2 gallon.
Carboy or other dispensing product sits upon a 16 x 12"More Product Information
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty