Datalogging Thermometer, Bluetooth 4-Channel, Sper Scientific
Supplier: Sper Scientific
800025
76214-010EA 610.73 USD
76214-010
Datalogging Thermometer, Bluetooth 4-Channel, Sper Scientific
Data Loggers
Wirelessly display, log and download to your computer or phone.
  • Bluetooth
  • High/low alarms
  • Fast response with high accuracy
  • Δ(T1 - T2)
  • Min./max./ave./hold
  • K, J, E, T, N, R, or S
  • Five year warranty
This unique portable thermometer displays data from 4 K,J,E,T,N,R, or S type thermocouple probes simultaneously on a large multi-line and back-lit LCD. Automatically record and store up to 32000 data points on each of the four channels.
Wireless bluetooth transmission eliminates the need to remove the thermometer to computer to download data. PreviouslyMore Product Information