Multi-Tube Vortex Mixer, G-Biosciences
Supplier: G-Biosciences
BT931
76263-870EA 2259.6 USD InStock
76263-870
Multi-Tube Vortex Mixer, G-Biosciences
Shakers and Mixers Vortex Mixers
Multi-tube vortex mixer is designed for mechanical and chemical cell lysis, mixing suspensions, and general sample agitation.
  • Microprocessor control, simple design, LED displays speed and time
  • User friendly program design, built-in short mix and timing operation modes. Operation is stable and silent
  • Large orbital vortex, effective performance
  • Multiple optional foam tube frames and pads are available for improved versatility
  • Can process 50 samples at the same time to make experiments more convenient and efficient
It gives a distinctly different motionMore Product Information
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty