Anti-V5 Tag Rabbit Monoclonal Antibody [clone: SV5-P-K]

Supplier: Enzo Life Sciences
ABS589-0200
76284-930EA 622 USD
76284-930
Anti-V5 Tag Rabbit Monoclonal Antibody [clone: SV5-P-K]
Antibodies
Recombinant monclonal antibody (Rabbit IgGκ).

Recommended Applications: ELISA, IF, IHC (FS), IP, WB, Radioimmunoassay

Type: Primary
Antigen: V5 Tag
Clonality: Monoclonal
Clone: SV5-P-K
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgGκ
Reactivity: Virus
Order Now