Humidity Chambers, Memmert
Supplier: Memmert
HCP50 HCP105 HCP150 HCP240
76299-472EA 13237.06 USD InStock
76299-472 76299-474 76299-476 76299-478
Humidity Chambers, Memmert
Testing Chambers Humidity Chambers
Newly upgraded features for easy use - still best for high humidity climate testing.
  • From temperatures of at least 7 °C above ambient up to +90 °C
  • Humidity range 20 to 95% RH or RH off
  • Programmable with internal data logging via included AtmoCONTROL software
  • In four sizes: 56 to 241 L volume chambers
From zoology and botany to material research, electronics and building physics: Environment simulations in the humidity chamber must be reliable and reproducible in the laboratory and be completely documentable. The high-precision temperature and humidityMore Product Information
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty